Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Carbenicillin disodium salt, free flowing powder

Supplier: E. Becker & Co. GmbH
Danger

02195092.2 0219509201 0219509205
IC195092.2EA 74.86 USD InStock
IC195092.2 IC19509201 IC19509205
Carbenicillin disodium salt, free flowing powder
Carbenicillin disodium salt
Semi-synthetic antibiotic related to penicillin. It is a benzyl penicillin derivative with substitution of a-carboxyl group n the benzyl side chain. The product interferes with final cell wall synthesis of susceptible bacteria. It is completely ionized in the blood. Carbencillin disodium is about 50% reversibly protein bound in plasma and its tissue distribution is similar to other penicillins.

  • Presentation: Free Flowing Powder
  • pKa: (in water) 3.3
  • Soluble in water (50 mg/ ml) and alcohol

Though Carbencillin disodium has substantial in vitro activity against a variety of both gram-positive and gram-negative microorganisms, the most important aspect of its profile is in its antipseudomonal and antiproteal effect.

Used for selection of ampr transformed cells and to study the role of penicillin-sensitive transpeptidases in cell wall biosynthesis.

Carboxypenicillin antibiotic that inhibits bacterial cell-wall synthesis (peptidoglycan cross-linking) by inactivating transpeptidases on the inner surface of the bacterial cell membrane. Analog to ampicillin. Antimicrobial spectrum: Gram-positive and Gram-negative bacteria, Pseudomonas.


Formula: C₁₇H₁₆N₂O₆S·2Na
MW: 422.363 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00077683
CAS Number: 4800-94-6
Order Now
Specification Test Results
Identity Test Passes
Potency >650 µg/mg

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty