Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-2-chloroacetoacetate ≥95.0%

Supplier: Decon Laboratories Ltd

C1219-25G C1219-250G
TCC1219-025GEA 76 USD InStock
TCC1219-025G TCC1219-250G
Methyl-2-chloroacetoacetate ≥95.0%
Methyl-2-chloroacetoacetate
CAS Number: 4755-81-1
MDL Number: MFCD00008757
Molecular Formula: C5H7ClO3
Molecular Weight: 150.56
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 86
Flash Point (°C): 71
Specific Gravity (20/20): 1.25

Other Product Names / Synonyms: 2-Chloroacetoacetic Acid Methyl Ester


Formula: C₅H₇ClO₃
MW: 150.56 g/mol
Boiling Pt: 137
Melting Pt: -33
Density: 1.25 (20 °C)
Flash Pt: 71 °C
MDL Number: MFCD00008757
CAS Number: 4755-81-1
EINECS: 225-286-6
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2eb5505b480983d11758aaef956b9002dacca631&vid=2eb5505b480983d11758aaef956b9002dacca631&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >95.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty