Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dodecylbenzenesulfonic acid mixture ≥95.0% (by titrimetric analysis), soft type

Supplier: Decon Laboratories Ltd
Danger

D0989-25G D0989-500G
TCD0989-025GEA 22.39 USD InStock
TCD0989-025G TCD0989-500G
Dodecylbenzenesulfonic acid mixture ≥95.0% (by titrimetric analysis), soft type
Dodecylbenzenesulphonic acid
CAS Number: 68584-22-5 MDL Number: MFCD00066677 Purity/Analysis Method: 95.0% (T) Form: Clear Liquid Flash Point (°C): 211

Other Product Names / Synonyms: Alkylbenzenesulfonic Acid, Laurylbenzenesulfonic Acid


Formula: C₁₈H₃₀O₃S
Boiling Pt: 315 °C (1013 hPa)
Melting Pt: 10 °C
Density: solvent 0.9 g/cm³ (25 °C)
Flash Pt: 211 °C
MDL Number: MFCD00066677
CAS Number: 27176-87-0
EINECS: 248-289-4
UN: 2586
ADR: 8,III
Order Now
Specification Test Results
Appearance Orange to Brown to Dark red clear liquid
Purity(Phase splitting method) min. 95.0 %
Neutralization Value 177.0 to 186.0
Petroleum ether soluble matter max. 2.5 %

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty