Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium-2-mercaptobenzothiazole ≥97.0%

Supplier: Decon Laboratories Ltd

M0057-25G M0057-500G
TCM0057-025GEA 35.1 USD InStock
TCM0057-025G TCM0057-500G
Sodium-2-mercaptobenzothiazole ≥97.0%
Sodium-2-mercaptobenzothiazole
CAS Number: 2492-26-4
MDL Number: MFCD00065334
Molecular Formula: C7H5NS2
Molecular Weight: 189.23
Purity/Analysis Method: >97.0% (HPLC)
Form: Crystal

Other Product Names / Synonyms: Sodium 2-Benzothiazolethiolate, 2-Mercaptobenzothiazole Sodium Salt


Formula: C₇H₅NS₂
MW: 189.23 g/mol
MDL Number: MFCD00065334
CAS Number: 2492-26-4
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d643df3aaaec4af382f8fa788190bc354eb6610b&vid=d643df3aaaec4af382f8fa788190bc354eb6610b&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >97.0% (HPLC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty