Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Mesityl oxide ≥95.0%

Supplier: Decon Laboratories Ltd
Danger

M0069-500ML M0069-25ML
TCM0069-500MLEA 187 USD InStock
TCM0069-500ML TCM0069-25ML
Mesityl oxide ≥95.0%
Mesityl oxide
CAS Number: 141-79-7
MDL Number: MFCD00008900
Molecular Formula: C6H10O
Molecular Weight: 98.15
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 130
Flash Point (°C): 27
Specific Gravity (20/20): 0.86

Other Product Names / Synonyms: Isopropylideneacetone, Mesityl Oxide


Formula: C₆H₁₀O
Boiling Pt: 130 °C
Density: 0.86 (20 °C)
Flash Pt: 27 °C
Storage Temperature: Ambient
MDL Number: MFCD00008900
CAS Number: 141-79-7
EINECS: 205-502-5
UN: 1229
ADR: 3,III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=93580d427cfd75c86ffc70d8132be7d0b39ada45&vid=93580d427cfd75c86ffc70d8132be7d0b39ada45&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >95.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty