Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-CD44 Rat Monoclonal Antibody [clone: IM7] (mFluor™ Violet)

Supplier: AAT BIOQUEST INC
10441101 10441100
77023-940EA 915.16 USD InStock
77023-940 77023-540
Anti-CD44 Rat Monoclonal Antibody [clone: IM7] (mFluor™ Violet)
Antibodies
IM7 is an anti-human/mouse monoclonal antibody that targets the CD44 antigen.

  • Purified through affinity chromatography

Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.

Precaución: For research use only.

Type: Secondary
Antigen: CD44
Clonality: Monoclonal
Clone: IM7
Conjugation: mFluor™ Violet
Epitope:
Host: Rat
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty