Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-CD161 Mouse Monoclonal Antibody [clone: HP-3G10] (iFluor® 660)

Supplier: AAT BIOQUEST INC
116100G0 116100G1
77022-416EA 244.24 USD InStock
77022-416 77023-474
Anti-CD161 Mouse Monoclonal Antibody [clone: HP-3G10] (iFluor® 660)
Antibodies
HP-3G10 is an anti-human monoclonal antibody that recognizes the CD161 antigen.

  • Purified through affinity chromatography

Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.

Precaución: For research use only.

Type: Secondary
Antigen: CD161
Clonality: Monoclonal
Clone: HP-3G10
Conjugation: iFluor® 660
Epitope:
Host: Mouse
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty