Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-CD3e Mouse Monoclonal Antibody [clone: SK7] (iFluor® 568)

Supplier: AAT BIOQUEST INC
100330B0 100330B1
77023-900EA 244.24 USD InStock
77023-900 77024-698
Anti-CD3e Mouse Monoclonal Antibody [clone: SK7] (iFluor® 568)
Antibodies
The SK7 monoclonal antibody reacts with human CD3e, a 20 kD member of the Ig superfamily often found on the surface of t cells, tregs and nkt cells.

  • Purified through affinity chromatography

Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.

Precaución: For research use only.

Type: Secondary
Antigen: CD3e
Clonality: Monoclonal
Clone: SK7
Conjugation: iFluor® 568
Epitope:
Host: Mouse
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty