Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-CD4 Mouse Monoclonal Antibody [clone: RPA-T4] (iFluor® 405)

Supplier: AAT BIOQUEST INC
10041020 10041021
77022-196EA 244.24 USD InStock
77022-196 77024-568
Anti-CD4 Mouse Monoclonal Antibody [clone: RPA-T4] (iFluor® 405)
Antibodies
RPA-T4 is an anti-human monoclonal antibody that targets the CD4 antigen.

  • Purified through affinity chromatography

Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.

Precaución: For research use only.

Type: Secondary
Antigen: CD4
Clonality: Monoclonal
Clone: RPA-T4
Conjugation: iFluor® 405
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty