Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Citric acid, anhydrous

Supplier: Bioanalytic
Warning

C34500-1000.0 C34500-500.0 C34500-5000.0 C34500-10000.0 C34500-25000.0
76345-410EA 65.99 USD InStock
76345-410 76345-412 76345-414 77510-206 77510-208
Citric acid, anhydrous
Citric acid
Precaución: Research or further manufacturing use only, not for food or drug use.


Formula: HOC(COOH)(CH₂COOH)₂
MW: 192.13 g/mol
Boiling Pt: 310
Melting Pt: 153...159 °C
Storage Temperature: Room Temperature
MDL Number: MFCD00011669
CAS Number: 77-92-9
Merck Index: 13,02350
Order Now
Specification Test Results
Appearance Fine white crystalline powder
Assay (Titration) ≥99.5 %
Carbonizable Substances (H2SO4) Pass Test
Clarity and Color Pass Test
Identification Pass Test
Lead ≤0.5 mg/kg
Limit of Oxalic Acid ≤0.036 %
Oxalate Test Pass Test
Residual Solvents Meet requirements
Residue on Ignition ≤0.05 %
Sulfate ≤0.015 %
Water ≤0.5 %
BSE/TSE Free BSE/TSE Free
Country of Origin Report

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty