Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dichlorobis(2,2'-bipyridine)ruthenium(II) ≥99%

Supplier: AMBEED, INC
Warning

A1329819-1G A1329819-250MG
77549-594EA 102 USD InStock
77549-594 77621-572
Dichlorobis(2,2'-bipyridine)ruthenium(II) ≥99%
Dichlorobis(2,2'-bipyridine)ruthenium(II)
Precaución: for Research Use Only


Formula: C₂₀H₁₆Cl₂N₄Ru
MDL Number: MFCD00792194
CAS Number: 15746-57-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty