Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

[[3,6-Dimethoxy-2',4',6'-tris(1-methylethyl)[1,1'-biphenyl]-2-yl-κC1']bis(1,1-dimethylethyl)phosphine-κP](η3-2-propen-1-yl)-Palladium(1+), 1,1,1-trifluoromethanesulfonate (1:1) ≥97%

Supplier: AMBEED, INC
Danger

A1360919-1G A1360919-100MG A1360919-250MG
77549-652EA 743 USD InStock
77549-652 77621-744 77621-746
[[3,6-Dimethoxy-2',4',6'-tris(1-methylethyl)[1,1'-biphenyl]-2-yl-κC1']bis(1,1-dimethylethyl)phosphine-κP](η3-2-propen-1-yl)-Palladium(1+), 1,1,1-trifluoromethanesulfonate (1:1) ≥97%
[[3,6-Dimethoxy-2',4',6'-tris(1-methylethyl)[1,1'-biphenyl]-2-yl-κC1']bis(1,1-dimethylethyl)phosphine-κP](η3-2-propen-1-yl)-Palladium(1+), 1,1,1-trifluoromethanesulfonate (1:1)
Precaución: for Research Use Only


Formula: C₃₄H₅₄O₂PPd·CF₃O₃S
CAS Number: 1798782-15-6
UN: 1759
ADR: 8,Ⅲ
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty