Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(5-Amino-1-oxoisoindolin-2-yl)piperidine-2,6-dione ≥97%

Supplier: AMBEED, INC
Danger

A1442988-100MG A1442988-250MG A1442988-50MG A1442988-5G
77622-102EA 151.41 USD InStock
77622-102 77622-104 77622-106 77622-108
3-(5-Amino-1-oxoisoindolin-2-yl)piperidine-2,6-dione ≥97%
3-(5-Amino-1-oxoisoindolin-2-yl)piperidine-2,6-dione
Precaución: for Research Use Only


Formula: C₁₃H₁₃N₃O₃
MDL Number: MFCD31692701
CAS Number: 191732-70-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty