Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Daunorubicin hydrochloride ≥98%

Supplier: AMBEED, INC
Danger

A148164-250MG A148164-100MG A148164-1G A148164-10MG
77549-822EA 58 USD InStock
77549-822 77615-188 77615-190 77685-244
Daunorubicin hydrochloride ≥98%
Daunorubicin hydrochloride
Precaución: for Research Use Only


Formula: C₂₇H₂₉NO₁₁·HCl
MDL Number: MFCD04974507
CAS Number: 23541-50-6
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty