Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fumaric acid monomethyl ester 98%

Supplier: AMBEED, INC
Warning

A251448-1000G A251448-500G
77668-058EA 63 USD InStock
77668-058 77668-060
Fumaric acid monomethyl ester 98%
Fumaric acid monomethyl ester
Precaución: For research use only.


Formula: C₅H₆O₄
MW: 130.1 g/mol
MDL Number: MFCD00063174
CAS Number: 2756-87-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty