Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1S,3R)-3-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid ≥97%

Supplier: AMBEED, INC
Warning

A358134-250MG A358134-25G
77672-796EA 22 USD InStock
77672-796 77672-798
(1S,3R)-3-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid ≥97%
(1S,3R)-3-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid
Precaución: for Research Use Only


Formula: C₁₂H₂₁NO₄
MW: 243.30 g/mol
MDL Number: MFCD06227743
CAS Number: 222530-34-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty