Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Rink Amide MBHA resin 0.3-0.8 mmol/g, resin 100-200 mesh, 1% DVB

Supplier: AMBEED, INC
Warning

A451407-100G A451407-1G
77673-116EA 422 USD InStock
77673-116 77673-118
Rink Amide MBHA resin 0.3-0.8 mmol/g, resin 100-200 mesh, 1% DVB
Rink Amide MBHA resin
Precaución: for Research Use Only


MDL Number: MFCD00165781
CAS Number: 431041-83-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty