Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Phytanic acid ≥95%

Supplier: AMBEED, INC
Warning

A617473-25G A617473-5G
77669-656EA 1668 USD InStock
77669-656 77669-658
Phytanic acid ≥95%
Phytanic acid
Precaución: for Research Use Only


Formula: C₂₀H₄₀O₂
MW: 312.53 g/mol
MDL Number: MFCD00133772
CAS Number: 14721-66-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty