Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-(-)-Tetrahydrofurfurylamine ≥98%

Supplier: AMBEED, INC
Danger

A628188-1G A628188-250MG
77551-890EA 32 USD InStock
77551-890 77747-293
(R)-(-)-Tetrahydrofurfurylamine ≥98%
(R)-(-)-Tetrahydrofurfurylamine
Precaución: for Research Use Only


Formula: C₅H₁₁NO
MDL Number: MFCD00192476
CAS Number: 7202-43-9
UN: 1993
ADR: 3,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty