Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N,N'-Dimethyl-1,3-propanediamine ≥98%

Supplier: AMBEED, INC
Danger

A676384-25G A676384-1G
77552-060EA 75 USD InStock
77552-060 77595-030
N,N'-Dimethyl-1,3-propanediamine ≥98%
N,N'-Dimethyl-1,3-propanediamine
Precaución: for Research Use Only


Formula: C₅H₁₄N₂
MW: 102.18 g/mol
MDL Number: MFCD00008292
CAS Number: 111-33-1
UN: 2733
ADR: 3,8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty