Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Selpercatinib 98%

Supplier: AMBEED, INC
Warning

A1145218-1G A1145218-1MG
77619-986EA 369.77 USD InStock
77619-986 77619-988
Selpercatinib 98%
Selpercatinib
Precaución: For research use only.


Formula: C₂₉H₃₁N₇O₃
MDL Number: MFCD31814089
CAS Number: 2152628-33-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty