Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

[2'-(Dicyclohexylphosphino-κP)-N2,N2,N6,N6-tetramethyl[1,1'-biphenyl]-2,6-diamine](methanesulfonato-κO)[2'-(methylamino-κN)[1,1'-biphenyl]-2-yl-κC]palladium 98%

Supplier: AMBEED, INC
Warning

A1155295-250MG A1155295-5G
77570-928EA 91 USD InStock
77570-928 77620-216
[2'-(Dicyclohexylphosphino-κP)-N2,N2,N6,N6-tetramethyl[1,1'-biphenyl]-2,6-diamine](methanesulfonato-κO)[2'-(methylamino-κN)[1,1'-biphenyl]-2-yl-κC]palladium 98%
[2'-(Dicyclohexylphosphino-κP)-N2,N2,N6,N6-tetramethyl[1,1'-biphenyl]-2,6-diamine](methanesulfonato-κO)[2'-(methylamino-κN)[1,1'-biphenyl]-2-yl-κC]palladium
Precaución: For research use only.


Formula: C₄₂H₅₆N₃O₃PPdS
MDL Number: MFCD30724404
CAS Number: 1810068-32-6
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty