Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Pyridin-2-yldisulfanyl)butanoic acid 97%

Supplier: AMBEED, INC
Warning

A1155653-100MG A1155653-250MG
77620-240EA 146.26 USD InStock
77620-240 77620-242
4-(Pyridin-2-yldisulfanyl)butanoic acid 97%
4-(Pyridin-2-yldisulfanyl)butanoic acid
Precaución: For research use only.


Formula: C₉H₁₁NO₂S₂
MDL Number: MFCD25961520
CAS Number: 250266-79-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty