Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-1-methyl-1,3-dihydro-2H-indol-2-one 96%

Supplier: AMBEED, INC
Danger

A148054-1G A148054-5G
77567-054EA 28 USD InStock
77567-054 77624-338
5-Bromo-1-methyl-1,3-dihydro-2H-indol-2-one 96%
5-Bromo-1-methyl-1,3-dihydro-2H-indol-2-one
Precaución: For research use only.


Formula: C₉H₈BrNO
MDL Number: MFCD00464061
CAS Number: 20870-90-0
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty