Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 5-bromo-4-oxopentanoate 98%, Ambeed.Inc

Supplier: AMBEED, INC
Danger

A386690-100MG A386690-5G
77684-113EA 19 USD InStock
77684-113 77684-114
Methyl 5-bromo-4-oxopentanoate 98%, Ambeed.Inc
Methyl 5-bromo-4-oxopentanoate
Precaución: For research use only.


Formula: C₆H₉BrO₃
MW: 209.04 g/mol
MDL Number: MFCD18800850
CAS Number: 53856-93-2
UN: 3265
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty