Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

BG-NH2 96%

Supplier: AMBEED, INC
Warning

A455042-100MG A455042-250MG
77568-892EA 47 USD InStock
77568-892 77673-142
BG-NH2 96%
BG-NH2
Precaución: For research use only.


Formula: C₁₃H₁₄N₆O
MDL Number: MFCD22577247
CAS Number: 674799-96-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty