Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 7-aminoheptanoate 95%

Supplier: AMBEED, INC
Warning

A838378-25G A838378-5G
77623-012EA 1180.38 USD InStock
77623-012 77623-014
tert-Butyl 7-aminoheptanoate 95%
tert-Butyl 7-aminoheptanoate
Precaución: For research use only.


Formula: C₁₁H₂₃NO₂
MW: 201.306 g/mol
MDL Number: MFCD12804784
CAS Number: 105974-64-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty