Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Fmoc-3-iodo-L-alanine methyl ester 98%

Supplier: AMBEED, INC
Warning

A888548-5G A888548-250MG
77573-794EA 72 USD InStock
77573-794 77749-695
N-Fmoc-3-iodo-L-alanine methyl ester 98%
N-Fmoc-3-iodo-L-alanine methyl ester
Precaución: For research use only.


Formula: C₁₉H₁₈INO₄
MDL Number: MFCD13189980
CAS Number: 156017-42-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty