Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3,5,6-Tetrafluorobenzenesulfonyl chloride 97%

Supplier: AMBEED, INC
Danger

A972971-100MG A972971-250MG
77749-810EA 53 USD InStock
77749-810 77749-811
2,3,5,6-Tetrafluorobenzenesulfonyl chloride 97%
2,3,5,6-Tetrafluorobenzenesulfonyl chloride
Precaución: For research use only.


Formula: C₆HClF₄O₂S
MDL Number: MFCD06409089
CAS Number: 776-09-0
UN: 3261
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty