Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 2-cyclopropyl-4-(4-fluorophenyl)quinoline-3-carboxylate ≥97%

Supplier: AMBEED, INC
Warning

A106500-10G A106500-5G
77670-522EA 62 USD InStock
77670-522 77670-524
Ethyl 2-cyclopropyl-4-(4-fluorophenyl)quinoline-3-carboxylate ≥97%
Ethyl 2-cyclopropyl-4-(4-fluorophenyl)quinoline-3-carboxylate
Precaución: for Research Use Only


Formula: C₂₁H₁₈FNO₂
MDL Number: MFCD09032923
CAS Number: 148516-11-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty