Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Moxidectin ≥98%

Supplier: AMBEED, INC
Danger

A109257-250MG A109257-100MG
77599-574EA 15.38 USD InStock
77599-574 77666-592
Moxidectin ≥98%
Moxidectin
Precaución: for Research Use Only


Formula: C₃₇H₅₃NO₈
MDL Number: MFCD00866560
CAS Number: 113507-06-5
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty