Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

PRE-084 hydrochloride ≥98%

Supplier: AMBEED, INC
Warning

A111935-10MG A111935-5MG
77745-828EA 76 USD InStock
77745-828 77745-829
PRE-084 hydrochloride ≥98%
PRE-084 hydrochloride
Precaución: for Research Use Only


Formula: C₁₉H₂₈ClNO₃
MDL Number: MFCD00153919
CAS Number: 75136-54-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty