Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ginsenoside Rd ≥98%

Supplier: AMBEED, INC
Warning

A114690-10MG A114690-100MG A114690-250MG
77614-898EA 8.24 USD InStock
77614-898 77747-833 77747-834
Ginsenoside Rd ≥98%
Ginsenoside Rd
Precaución: for Research Use Only


Formula: C₄₈H₈₂O₁₈
MDL Number: MFCD00210508
CAS Number: 52705-93-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty