Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-tert-Butyl 2-(2-hydroxyethyl)piperazine-1-carboxylate ≥95%

Supplier: AMBEED, INC
Warning

A114817-1G A114817-250MG A114817-100MG
77747-837EA 322 USD InStock
77747-837 77747-838 77747-836
(R)-tert-Butyl 2-(2-hydroxyethyl)piperazine-1-carboxylate ≥95%
(R)-tert-Butyl 2-(2-hydroxyethyl)piperazine-1-carboxylate
Precaución: for Research Use Only


Formula: C₁₁H₂₂N₂O₃
MDL Number: MFCD12756015
CAS Number: 947275-74-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty