Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2R)-2,3-Bis((3,7,11,15-tetramethylhexadecanoyl)oxy)propyl (2-(trimethylammonio)ethyl) phosphate ≥99%

Supplier: AMBEED, INC
Danger

A1152769-100MG A1152769-250MG A1152769-50MG
77620-152EA 215.27 USD InStock
77620-152 77620-154 77620-156
(2R)-2,3-Bis((3,7,11,15-tetramethylhexadecanoyl)oxy)propyl (2-(trimethylammonio)ethyl) phosphate ≥99%
(2R)-2,3-Bis((3,7,11,15-tetramethylhexadecanoyl)oxy)propyl (2-(trimethylammonio)ethyl) phosphate
Precaución: for Research Use Only


Formula: C₄₈H₉₆NO₈P
MW: 846.2515 g/mol
MDL Number: MFCD00674308
CAS Number: 207131-40-6
UN: 2811
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty