Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Deforolimus ≥98%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A116842-10MG A116842-5MG
77683-054EA 99 USD InStock
77683-054 77683-055
Deforolimus ≥98%, Ambeed.Inc
Deforolimus
Precaución: For research use only.


Formula: C₅₃H₈₄NO₁₄P
MDL Number: MFCD13184816
CAS Number: 572924-54-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty