Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

m-PEG12-NHS ester ≥98%

Supplier: AMBEED, INC
Warning

A1174640-100MG A1174640-1G A1174640-250MG
77620-422EA 86.52 USD InStock
77620-422 77620-424 77620-426
m-PEG12-NHS ester ≥98%
m-PEG12-NHS ester
Precaución: for Research Use Only


Formula: C₃₀H₅₅NO₁₆
MDL Number: MFCD13184970
CAS Number: 2207596-93-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty