Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-5-(5-(1-(2,6-Dichloro-3-fluorophenyl)ethoxy)pyridin-3-yl)pyrimidin-2-amine 98+%

Supplier: AMBEED, INC

A1176897-100MG A1176897-25MG A1176897-50MG
77620-538EA 310.03 USD InStock
77620-538 77620-540 77620-542
(R)-5-(5-(1-(2,6-Dichloro-3-fluorophenyl)ethoxy)pyridin-3-yl)pyrimidin-2-amine 98+%
(R)-5-(5-(1-(2,6-Dichloro-3-fluorophenyl)ethoxy)pyridin-3-yl)pyrimidin-2-amine
Precaución: for Research Use Only


Formula: C₁₇H₁₃Cl₂FN₄O
MDL Number: MFCD31813691
CAS Number: 2252247-80-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty