Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-((2-Aminoethyl)disulfanyl)propanoic acid ≥95%

Supplier: AMBEED, INC

A119413-100MG A119413-1G A119413-250MG A119413-50MG
77670-688EA 154 USD InStock
77670-688 77670-690 77670-692 77670-694
3-((2-Aminoethyl)disulfanyl)propanoic acid ≥95%
3-((2-Aminoethyl)disulfanyl)propanoic acid
Precaución: for Research Use Only


Formula: C₅H₁₁NO₂S₂
MDL Number: MFCD07357249
CAS Number: 15579-00-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty