Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Carbonyldiiodo(pentamethylcyclopentadienyl)cobalt(III) ≥95%

Supplier: AMBEED, INC

A1195873-250MG A1195873-5G
77620-762EA 25.75 USD InStock
77620-762 77620-764
Carbonyldiiodo(pentamethylcyclopentadienyl)cobalt(III) ≥95%
Carbonyldiiodo(pentamethylcyclopentadienyl)cobalt(III)
Precaución: for Research Use Only


Formula: C₁₁H₁₅CoI₂O
MW: 475.98 g/mol
MDL Number: MFCD00013702
CAS Number: 35886-64-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty