Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(2-(4-Aminopiperidin-1-yl)-5-(3-fluoro-4-methoxyphenyl)-1-methyl-6-oxo-1,6-dihydropyrimidin-4-yl)-2-fluorobenzonitrile benzenesulfonate ≥97%

Supplier: AMBEED, INC

A1207394-100MG A1207394-50MG
77620-818EA 394.49 USD InStock
77620-818 77620-820
4-(2-(4-Aminopiperidin-1-yl)-5-(3-fluoro-4-methoxyphenyl)-1-methyl-6-oxo-1,6-dihydropyrimidin-4-yl)-2-fluorobenzonitrile benzenesulfonate ≥97%
4-(2-(4-Aminopiperidin-1-yl)-5-(3-fluoro-4-methoxyphenyl)-1-methyl-6-oxo-1,6-dihydropyrimidin-4-yl)-2-fluorobenzonitrile benzenesulfonate
Precaución: for Research Use Only


Formula: C₃₀H₂₉F₂N₅O₅S
MW: 609.64 g/mol
MDL Number: MFCD32644589
CAS Number: 2097523-60-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty