Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(7-Hydroxy-1-oxoisoindolin-2-yl)piperidine-2,6-dione ≥97%

Supplier: AMBEED, INC

A1213292-100MG A1213292-250MG A1213292-50MG
77620-910EA 71.07 USD InStock
77620-910 77620-912 77620-914
3-(7-Hydroxy-1-oxoisoindolin-2-yl)piperidine-2,6-dione ≥97%
3-(7-Hydroxy-1-oxoisoindolin-2-yl)piperidine-2,6-dione
Precaución: for Research Use Only


Formula: C₁₃H₁₂N₂O₄
MW: 260.25 g/mol
CAS Number: 1416990-10-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty