Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N6-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-fluoro-2'-deoxyadenosine-3'-(2-cyanoethyl diisopropylphosphoramidite) ≥95%

Supplier: AMBEED, INC
Warning

A122283-100MG A122283-1G A122283-5G
77685-079EA 18 USD InStock
77685-079 77685-080 77685-081
N6-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-fluoro-2'-deoxyadenosine-3'-(2-cyanoethyl diisopropylphosphoramidite) ≥95%
N6-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-fluoro-2'-deoxyadenosine-3'-(2-cyanoethyl diisopropylphosphoramidite)
Precaución: for Research Use Only


Formula: C₄₇H₅₁FN₇O₇P
MDL Number: MFCD12912392
CAS Number: 136834-22-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty