Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Endo-bcn-peg2-acid ≥98%

Supplier: AMBEED, INC

A1225965-100MG A1225965-1G A1225965-250MG A1225965-50MG
77621-126EA 337.84 USD InStock
77621-126 77621-128 77621-130 77621-132
Endo-bcn-peg2-acid ≥98%
Endo-bcn-peg2-acid
Precaución: for Research Use Only


Formula: C₁₈H₂₇NO₆
MW: 353.41 g/mol
MDL Number: MFCD31692189
CAS Number: 1993134-72-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty