Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-5-(3-oxo-4-(2-(trifluoromethyl)benzyl)piperazin-1-yl)pyridazin-3(2H)-one ≥98%

Supplier: AMBEED, INC

A1226349-100MG A1226349-50MG
77621-140EA 176.13 USD InStock
77621-140 77621-142
4-Chloro-5-(3-oxo-4-(2-(trifluoromethyl)benzyl)piperazin-1-yl)pyridazin-3(2H)-one ≥98%
4-Chloro-5-(3-oxo-4-(2-(trifluoromethyl)benzyl)piperazin-1-yl)pyridazin-3(2H)-one
Precaución: for Research Use Only


Formula: C₁₆H₁₄ClF₃N₄O₂
MDL Number: MFCD32671553
CAS Number: 2304549-73-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty