Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(Z)-(4-((4-((3-(4-Fluorobenzyl)-2,4-dioxothiazolidin-5-ylidene)methyl)phenoxy)methyl)phenyl)boronic acid ≥98%

Supplier: AMBEED, INC

A1251838-10MG A1251838-5MG
77621-268EA 67.98 USD InStock
77621-268 77621-270
(Z)-(4-((4-((3-(4-Fluorobenzyl)-2,4-dioxothiazolidin-5-ylidene)methyl)phenoxy)methyl)phenyl)boronic acid ≥98%
(Z)-(4-((4-((3-(4-Fluorobenzyl)-2,4-dioxothiazolidin-5-ylidene)methyl)phenoxy)methyl)phenyl)boronic acid
Precaución: for Research Use Only


Formula: C₂₄H₁₉BFNO₅S
MW: 463.29 g/mol
CAS Number: 1312201-00-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty