Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dihydrolevoglucosenone ≥98%

Supplier: AMBEED, INC
Warning

A1255631-10G A1255631-100G A1255631-25G A1255631-5G
77596-594EA 28.7 USD InStock
77596-594 77621-298 77621-300 77621-302
Dihydrolevoglucosenone ≥98%
Dihydrolevoglucosenone
Precaución: for Research Use Only


Formula: C₆H₈O₃
MDL Number: MFCD23105760
CAS Number: 53716-82-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty