Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Hydrogen 3-(2-carboxylatoethyl)-5,5-difluoro-7-(1H-pyrrol-2-yl)-5H-dipyrrolo[1,2-c:2',1'-f][1,3,2]diazaborinin-4-ium-5-uide ≥98%

Supplier: AMBEED, INC

A1280331-250MG A1280331-50MG
77621-460EA 613.88 USD InStock
77621-460 77621-462
Hydrogen 3-(2-carboxylatoethyl)-5,5-difluoro-7-(1H-pyrrol-2-yl)-5H-dipyrrolo[1,2-c:2',1'-f][1,3,2]diazaborinin-4-ium-5-uide ≥98%
Hydrogen 3-(2-carboxylatoethyl)-5,5-difluoro-7-(1H-pyrrol-2-yl)-5H-dipyrrolo[1,2-c:2',1'-f][1,3,2]diazaborinin-4-ium-5-uide
Precaución: for Research Use Only


Formula: C₁₆H₁₄BF₂N₃O₂
CAS Number: 150173-78-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty