Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Methyl-1H-indole-2-carboxylic acid ≥95%

Supplier: AMBEED, INC
Warning

A131480-100MG A131480-1G A131480-250MG A131480-5G
77747-933EA 16 USD InStock
77747-933 77747-934 77747-935 77747-936
3-Methyl-1H-indole-2-carboxylic acid ≥95%
3-Methyl-1H-indole-2-carboxylic acid
Precaución: for Research Use Only


Formula: C₁₀H₉NO₂
MW: 175.18 g/mol
MDL Number: MFCD01652241
CAS Number: 10590-73-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty