Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-N-(4-((4-methyl-6-(phenylamino)pyrimidin-2-yl)amino)phenyl)benzamide ≥98%

Supplier: AMBEED, INC

A1339685-10MG A1339685-5MG
77621-598EA 116.39 USD InStock
77621-598 77621-600
2-Chloro-N-(4-((4-methyl-6-(phenylamino)pyrimidin-2-yl)amino)phenyl)benzamide ≥98%
2-Chloro-N-(4-((4-methyl-6-(phenylamino)pyrimidin-2-yl)amino)phenyl)benzamide
Precaución: for Research Use Only


Formula: C₂₄H₂₀ClN₅O
MW: 429.9 g/mol
MDL Number: MFCD09238311
CAS Number: 946293-78-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty